Learn More
C9orf78 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546104
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NRRDEDADMM KYIETELKKR KGIVEHEEQK VKPKNAEDCL YELPENIRVS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 100%; Zebrafish: 79%
C9orf78 Polyclonal antibody specifically detects C9orf78 in Human samples. It is validated for Western Blot.Spezifikation
C9orf78 | |
Polyclonal | |
Unconjugated | |
C9orf78 | |
bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZ63 | |
C9orf78 | |
Synthetic peptide directed towards the middle region of human C9orf78 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |