missing translation for 'onlineSavingsMsg'
Learn More

RABL6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531048

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13475557

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (95%), Rat (95%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RABL6 Polyclonal antibody specifically detects RABL6 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

RABL6
Polyclonal
Unconjugated
RABL6
B230208H17Rik; C11H9orf86; C9H9orf86; C9orf86; GTP-binding protein Parf; PARF; Partner of ARF; pp8875; putative GTP-binding protein Parf; RAB, member of RAS oncogene family-like 6; RAB, member RAS oncogene family like 6; RAB, member RAS oncogene family-like 6; RABL6; Rab-like GTP-binding protein 1; Rab-like GTP-binding protein 1A; Rab-like GTP-binding protein 1C; rab-like protein 1; Rab-like protein 6; RBEL1; Rbel1a; Rbel1b; RGD1307615
Rabbit
Antigen affinity chromatography
RUO
55684
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
0.77 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q3YEC7
RABL6
Recombinant fragment corresponding to a region within amino acids 42 and 302 of Human C9orf86
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote