missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18764
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Canine IgG (H+L) Polyclonal antibody specifically detects Canine IgG (H+L) in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
Canine IgG (H+L) | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Canine | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |