Learn More
CSNK1A1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528272
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, rat thymus. Predicted reactivity: Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CSNK1A1L Polyclonal antibody specifically detects CSNK1A1L in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
CSNK1A1L | |
Polyclonal | |
Unconjugated | |
CSNK1A1L | |
casein kinase 1 alpha 1 like; casein kinase 1, alpha 1-like; casein kinase I alpha S-like; casein kinase I isoform alpha-like; CK1; CKI-alpha-like; CSNK1A1L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
122011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8N752 | |
CSNK1A1L | |
Recombinant fragment corresponding to a region within amino acids 10 and 228 of Human Casein Kinase 1 alpha 1L | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |