missing translation for 'onlineSavingsMsg'
Learn More

Caspase 8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA513498

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 10309739

  • 514.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

Caspase 8 Polyclonal antibody specifically detects Caspase 8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Caspase 8
Polyclonal
PBS with 0.09% sodium azide
Q14790
Casp8
KLH conjugated synthetic peptide between 432-461 amino acids from the C-terminal region of human CASP8
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Casp8
ALPS2B; apoptosis-related cysteine peptidase; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; CASP8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; Caspase8; caspase-8; Caspase-8 precursor; Caspase-8 subunit p10; Caspase-8 subunit p18; EC 3.4.22.61; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; FLJ17672; ICE8; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MGC78473; MORT1-associated ced-3 homolog; OTTHUMP00000163720; OTTHUMP00000165063; OTTHUMP00000206557; OTTHUMP00000206581
Rabbit
Antigen affinity chromatography, Protein A
RUO
841
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers