Learn More
Caspase 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA137232
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta tissue.
Caspase 1 Polyclonal antibody specifically detects Caspase 1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Caspase 1 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P29466 | |
CASP1 | |
Synthetic peptide derived from internal region of human caspase 1 protein | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
CASP1 | |
CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
834 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |