Learn More
Caspase 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527387
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116, THP-1 conditioned medium (500 ng/mL LPS treatment for 4 hr and 1 mM ATP treatment for 1.5hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caspase 1 Polyclonal antibody specifically detects Caspase 1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Caspase 1 | |
Polyclonal | |
Unconjugated | |
CASP1 | |
CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.08 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P29466 | |
CASP1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 197 of Human Caspase 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |