missing translation for 'onlineSavingsMsg'
Learn More

Caspase 10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA519870

Additional Details : Netto-Gewicht : 0.01000kg

Product Code. 12878082

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

In Western blot applications, this antibody detects a band at ∽59kDa. A suggested positive control is Hela cell lysate.

Caspase 10 Polyclonal antibody specifically detects Caspase 10 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Caspase 10
Polyclonal
Unconjugated
CASP10
ALPS2; Apoptotic protease Mch-4; CASP10; CASP-10; caspase 10; caspase 10 apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine peptidase; caspase 10, apoptosis-related cysteine protease; caspase-10; Caspase-10 subunit p12; Caspase-10 subunit p23/17; FADD-like ICE2; FAS-associated death domain protein interleukin-1B-converting enzyme 2; FLICE2; ICE-like apoptotic protease 4; interleukin-1B-converting enzyme 2; MCH4
Rabbit
Ion-exchange chromatography
RUO
843
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q92851
CASP10
A 17 amino acid peptide near the carboxy terminus of human Caspase-10
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers