missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517802
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other caspases.
Caspase 12 Polyclonal antibody specifically detects Caspase 12 in Mouse samples. It is validated for Western Blot.Spezifikation
Caspase 12 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O08736 | |
CASP12 | |
Synthetic peptide corresponding to residues surrounding amino acid 158 of mouse caspase-12 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CASP12 | |
CASP12; CASP-12; CASP12P1; caspase 12; caspase 12 (gene/pseudogene); caspase 12 pseudogene 1; caspase-12; Inactive caspase-12; OTTMUSP00000045799; OTTMUSP00000045802; OTTMUSP00000045803; UNQ9415; UNQ9415/PRO34398 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12364 | |
-20°C | |
Liquid |