missing translation for 'onlineSavingsMsg'
Learn More

Caspase 12 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA112508

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11535992

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

A suggested positive control for this product is murine spleen tissue lysate.

Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
Spezifikation

Spezifikation

Caspase 12
Polyclonal
PBS with 0.02% sodium azide
Q6UXS9, O08736, Q920D5
Rabbit
Antigen affinity chromatography
RUO
100506742, 12364, 156117
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
Unconjugated
CASP12
CASP12
Synthetic peptide corresponding to residues 100-116 of the murine caspase-12.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote