Learn More
Caspase 2 p18 (Cleaved Thr325) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA539872
Weitere Details : Netto-Gewicht : 0.00900kg
Artikelnummer. 15516741
  • 452.00 € / 100 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Spezifikation
Spezifikation
Caspase 2 p18 (Cleaved Thr325)
Polyclonal
Unconjugated
CASP2
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57, caspase-2, neural precursor cell expressed developmentally down-regulated protein 2, protease ICH-1, protein phosphatase 1, regulatory subunit 57
Rabbit
Antigen affinity chromatography
RUO
12366, 64314, 835
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P42575, P29594, P55215
CASP2
A synthetic peptide derived from human CASP2
100 μg
Primary
Human, Mouse, Rat