Learn More
Caspase 3 Recombinant Rabbit Monoclonal Antibody (9H19L2), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 700182
Weitere Details : Netto-Gewicht : 1.00000kg
Beschreibung
This antibody is predicted to react with bovine, canine, feline, hamster, mouse, primate, pufferfish, rabbit, rat, porcine and Xenopus based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Caspase 3 Recombinant Monoclonal antibody specifically detects Caspase 3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Caspase 3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P42574, P70677 | |
CASP3 | |
A peptide corresponding to amino acids 171-175 of P42574. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
9H19L2 | |
Unconjugated | |
CASP3 | |
A830040C14Rik; AC-3; Apopain; Apopain precursor; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase3; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; Caspase-3-like protein; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; cysteine protease; CPP32; apopain; ICE3; IRP; LICE; mLdy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein | |
Rabbit | |
Protein A | |
RUO | |
12367, 836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |