missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534413
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-34413 can be used with blocking peptide PEP-1455.
Caspase 3 Polyclonal antibody specifically detects Caspase 3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
Caspase 3 | |
Polyclonal | |
Unconjugated | |
CASP3 | |
A830040C14Rik; AC-3; Apopain; Apopain precursor; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase3; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; Caspase-3-like protein; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; cysteine protease; CPP32; apopain; ICE3; IRP; LICE; mLdy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
836 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P42574 | |
CASP3 | |
A 17 amino acid synthetic peptide near the center of human Caspase-3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |