missing translation for 'onlineSavingsMsg'
Learn More

Caspase 6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA128386

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11515853

  • 451.00 € / 50 Mikrogramm
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-28386 detects Caspase 6 from hamster, rat, mouse, human, ovine, canine, rabbit, non-human primate samples. PA1-28386 has been successfully used in Western blot applications. The PA1-28386 immunogen is: Available on request.

Caspase 6 is a member of the cysteine-aspartic acid protease (caspase) family of enzymes. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic acid residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein is processed by caspases 7, 8 and 10, and is thought to function as a downstream enzyme in the caspase activation cascade. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Spezifikation

Spezifikation

Caspase 6
Polyclonal
Unconjugated
P55212, O08738, O35397
CASP6
Available on request.
50 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
0.2 mg/mL
CASP6
Mch2, mCASP-6
Rabbit
Antigen affinity chromatography
RUO
12368, 83584, 839
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote