Learn More
Caspase 7 Monoclonal Antibody (B4-G2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA191217
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-91217 detects Caspase 7 from mouse, rat, human samples. MA1-91217 has been successfully used in immunoprecipitation, Western blot applications. The MA1-91217 immunogen is: Available on request.
Caspase 7 is part of a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. The apoptotic subfamily can be further divided into initiator caspases, which are activated in response to death signals, and executioner caspases, which are activated by the initiator caspases and are responsible for cleavage of cellular substrates that ultimately lead to cell death. Caspase 7 is an executioner caspase that was identified based on its homology with caspases 1 and 3, as well as the C. elegans cell death protein CED-3. Alternative splicing of Caspase-7 mRNA results in the production of 3 distinct isoforms. Caspase-7 activity can be directly inhibited by XIAP expression. Alternative splicing results in four transcript variants, encoding three distinct isoforms of Caspase 7. Diseases associated with CASP7 include Type 1 Diabetes Mellitus 17 and Aleutian Mink Disease.Spezifikation
Caspase 7 | |
Monoclonal | |
0.5 mg/mL | |
CASP7 | |
AI314680, CMH-1, ICE-IAP3, Mch3, caspase-7, mCASP-7 | |
Mouse | |
Purified | |
RUO | |
12369, 64026, 840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
B4-G2 | |
Unconjugated | |
P55210, P97864 | |
CASP7 | |
Available on request. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |