Learn More
Caspase 7 Monoclonal Antibody (S.606.3), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515159
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other caspases.
Caspase 7 Monoclonal antibody specifically detects Caspase 7 in Human samples. It is validated for Western Blot.Spezifikation
Caspase 7 | |
Monoclonal | |
140 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P55210 | |
CASP7 | |
Recombinant human caspase-7 protein | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
S.606.3 | |
Unconjugated | |
CASP7 | |
AI314680; Apoptotic protease Mch-3; Casp7; CASP-7; caspase 7; caspase 7, apoptosis-related cysteine peptidase; caspase 7, apoptosis-related cysteine protease; caspase-7; Caspase-7 subunit p11; Caspase-7 subunit p20; CMH-1; cysteine protease LICE2; ICE-IAP3; ICE-LAP3; ICE-like apoptotic protease 3; Lice2; mCASP-7; MCH3; OTTHUMP00000020511; OTTHUMP00000020513; OTTHUMP00000020514; RP11-211N11.6 | |
Mouse | |
Affinity chromatography | |
RUO | |
840 | |
-20°C | |
Liquid |