Learn More
Caspase 8 Monoclonal Antibody (90A992), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA141280
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-41280 detects Caspase-8 in human, Rhesus monkey, and chimpanzee samples. Suggested positive control: Jurkat, antigen standard for CASP8 (transient overexpression lysate), Jurkat whole cell lysate.
Caspase 8 Monoclonal antibody specifically detects Caspase 8 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Caspase 8 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q14790 | |
Casp8 | |
Amino acids 360-385 of the human Caspase 8 protein. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
90A992 | |
Unconjugated | |
Casp8 | |
ALPS2B; apoptosis-related cysteine peptidase; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; CASP8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; Caspase8; caspase-8; Caspase-8 precursor; Caspase-8 subunit p10; Caspase-8 subunit p18; EC 3.4.22.61; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; FLJ17672; ICE8; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MGC78473; MORT1-associated ced-3 homolog; OTTHUMP00000163720; OTTHUMP00000165063; OTTHUMP00000206557; OTTHUMP00000206581 | |
Mouse | |
Protein G | |
RUO | |
841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |