missing translation for 'onlineSavingsMsg'
Learn More

Catenin alpha-1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA125081

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11585583

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: A431 and cytosolic fraction of rat embryonic brain, MDCK, MCF-7. PA1-25081 is expected to cross react with mouse (100% conserved) and Xenopus laevis (100% conserved) due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Catenin alpha-1 Polyclonal antibody specifically detects Catenin alpha-1 in Canine, Human, Mouse, Rat, Xenopus laevis samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Catenin alpha-1
Polyclonal
0.01M PBS with 15mM sodium azide; pH 7.4
P26231, P35221
CTNNA1, ctnna1.S
Synthetic peptide, conjugated to KLH, corresponding to residues E(873) K Q D E T Q T K I K R A S Q(887) of human alpha 1 Catenin.
100 μL
Primary
Canine, Human, Mouse, Rat, X. laevis
Antibody
IgG
Immunocytochemistry, Western Blot
Unconjugated
CTNNA1
[a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg
Rabbit
IgG fraction
RUO
12385, 1495, 307505, 399303, 474698
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote