Learn More
beta Catenin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA516429
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-16429 targets Catenin-beta in immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with mouse, rat, Xenopus, chicken, and human samples. The PA5-16429 immunogen is a synthetic peptide from the C-terminus of the beta-catenin protein.
beta Catenin Polyclonal antibody specifically detects beta Catenin in Chicken, Human, Mouse, Rat, X. laevis samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
beta Catenin | |
Polyclonal | |
Unconjugated | |
CTNNB1 | |
armadillo; armadillo homolog; Bcatenin; b-catenin; beta 1 88kDa; beta catenin; Betacatenin; beta-catenin; beta-catenin 1; Bfc; C20orf33; Cadherin associated protein; catenin; catenin (cadherin associated protein), beta 1; catenin (cadherin associated protein), beta 1, 88kDa; catenin (cadherin-associated protein) beta 1; catenin (cadherin-associated protein), beta 1; catenin (cadherin-associated protein), beta 1, 88kDa; Catenin b 1; Catenin b1; catenin beta; catenin beta 1; catenin beta 1 L homeolog; Catenin beta1; Catenin beta-1; Catenin β 1; Catenin β1; Catnb; CHBCAT; CTNB1; CTNNB; ctnnb1; ctnnb1.L; ctnnb1-a; ctnnb1-b; dJ633O20.1; DKFZp686D02253; FLJ25606; FLJ37923; id:ibd2058; Mesc; MRD19; NAP; NYD-SP19; OK/SW-cl.35; OTTHUMP00000209289; P14L; PP8304; PRO2286; RP5-1118M15.1; wu:fb73e10; wu:fi81c06; wu:fk25h01; XELAEV_18031149mg; β catenin; βcatenin | |
Rabbit | |
Protein A | |
RUO | |
12387, 1499, 395964, 399274, 84353 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P35222, Q02248, Q9WU82 | |
CTNNB1, ctnnb1.L | |
A synthetic peptide from the C-terminus of the beta-catenin protein | |
500 μL | |
Primary | |
Chicken, Human, Mouse, Rat, X. laevis | |
Antibody | |
IgG |