missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L2 (Cleaved Leu114) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539874
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Cathepsin V (CTSU, CTSV, CATL2, cathepsin U, cathepsin L2) is a lysosomal cysteine proteinase that influences corneal physiology, and mediates degradation of invariant chain in human thymus. Cathepsin V is present in corneal epithelium, activated macrophages, and colorectal and breast carcinomas. A 1.8-kb mRNA to Cathepsin V is present in normal human thymus and testis. Cathepsin V is a member of the peptidase C1 family. Cysteine proteinases are synthesized as preproenzymes, which are processed to the corresponding proenzymes. The proenzymes are either targeted to the lysosome or continue along the cellular secretory route.Spezifikation
Cathepsin L2 (Cleaved Leu114) | |
Polyclonal | |
Unconjugated | |
CTSV | |
Cathepsin L2, Cathepsin U, Cathepsin V, CTSV, CATL2, CTSL2, CTSU, UNQ268, PRO305, CATL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1515 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60911 | |
CTSV | |
A synthetic peptide derived from human CATL2 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |