missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530002
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Predicted reactivity: Dog (85%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cathepsin S Polyclonal antibody specifically detects Cathepsin S in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Cathepsin S | |
Polyclonal | |
Unconjugated | |
CTSS | |
Cat S; cathepsin S; Cats; CTSS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P25774 | |
CTSS | |
Recombinant protein fragment corresponding to a region within amino acids 143 and 320 of Human Cathepsin S | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |