missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CATSPERB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531585
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CATSPERB Polyclonal antibody specifically detects CATSPERB in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
CATSPERB | |
Polyclonal | |
Unconjugated | |
CATSPERB | |
C14orf161; cation channel sperm associated auxiliary subunit beta; cation channel sperm-associated protein subunit beta; cation channel, sperm associated beta; cation channel, sperm-associated, beta; catsper channel auxiliary subunit beta; CatSper(beta); CATSPERB; CatSper-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H7T0 | |
CATSPERB | |
Recombinant fragment corresponding to a region within amino acids 1 and 278 of Human CATSPERB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |