Learn More
Caveolin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532689
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Baboon, Monkey, Galago, Marmoset, Mouse, Rat, Ferret, Sheep, Bovine, Dog, Cat, Bat, Horse, Rabbit, Pig, Guinea pig (100%); Elephant, Armadillo (93%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Caveolin 1 Polyclonal antibody specifically detects Caveolin 1 in Bat, Bovine, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Caveolin 1 | |
Polyclonal | |
Unconjugated | |
CAV1 | |
BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100008837, 100055975, 100718512, 12389, 25404, 281040, 404693, 445468, 857 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P41350, P49817, P79132, Q03135, Q09YN6, Q2QLB0, Q6B3Y2, Q6RVA9 | |
CAV1 | |
Synthetic 15 amino acid peptide from N-terminus of human CAV1 / Caveolin. | |
50 μg | |
Primary | |
Bat, Rat, Bovine, Equine, Guinea Pig, Mouse, Mouse, Ovine, Ovine, Rabbit, Rabbit | |
Antibody | |
IgG |