Learn More
Caveolin 1 Monoclonal Antibody (7C8), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA140241
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-40241 detects Caveolin-1 from rat samples. MA1-40241 recognizes caveolin-1 alpha as well as caveolin-1 beta, which are present in many tissues, like aorta, heart, muscle, lung, adipose white, brown and epidydimal fat. MA1-40241 can be used to immuno-isolate caveolae. MA1-40241 has been successfully used in immunofluorescence, immunoprecipitation and Western blot applications. The MA1-40241 immunogen is rat caveolin. Positive control: Adipocytes (3T3-L1 adipocytes); Negative control: Cytoplasmic extracts of adipocytes.
Caveolin 1 Monoclonal antibody specifically detects Caveolin 1 in Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Caveolin 1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P41350 | |
CAV1 | |
Rat caveolin-1 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
7C8 | |
Unconjugated | |
CAV1 | |
BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04 | |
Mouse | |
Protein G | |
RUO | |
25404 | |
4° C | |
Liquid |