missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin 1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA19021
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology.
Caveolin 1 Polyclonal antibody specifically detects Caveolin 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Caveolin 1 | |
Polyclonal | |
Unconjugated | |
CAV1 | |
BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
857 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q03135 | |
CAV1 | |
Internal sequence corresponding to amino acids C-DELSEKQVYDAH. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |