missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517447
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with caveolin-2 or -3.
Caveolin 1 Polyclonal antibody specifically detects Caveolin 1 in Bovine, Chicken, Hamster, Human, Mouse, Porcine, Rat, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
Caveolin 1 | |
Polyclonal | |
Unconjugated | |
CAV1 | |
BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100753021, 12389, 25404, 281040, 323695, 373996, 404693, 857 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
33 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P35431, P41350, P49817, P79132, Q03135, Q6RVA9 | |
CAV1 | |
Synthetic peptide corresponding to residues surrounding Glu20 of human caveolin-1 | |
100 μL | |
Primary | |
Bovine, Chicken, Hamster, Human, Mouse, Porcine, Rat, Zebrafish | |
Antibody | |
IgG |