missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin 2 Monoclonal Antibody (ZC013), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 410700
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Caveolin 2 Monoclonal antibody specifically detects Caveolin 2 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
Caveolin 2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P51636 | |
CAV2 | |
Synthetic peptide derived from the C-terminal region of the human caveolin-2 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
ZC013 | |
Unconjugated | |
CAV2 | |
AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 | |
Mouse | |
Protein A | |
RUO | |
858 | |
-20°C | |
Liquid |