missing translation for 'onlineSavingsMsg'
Learn More

CD11a Mouse anti-Human, Unlabeled, Clone: 27, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610826

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15895659

  • 308.94 € / 1 Set
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD11a, also known as integrin αL or LFA-1 (lymphocyte function-associated antigen-1), is a member of the β2 family of integrins that includes Mac-1 and p150,95. It is composed of two noncovalently linked α and β chains which are associated with the actin cytoskeleton. CD11a (integrin αL) has been reported to be expressed on the surface of T cells, neutrophils, and macrophages. It is involved in cell-cell adhesion and it also conveys signals from the plasma membrane to the cytoplasm. Activation of the TCR/CD3 complex on T cells transmits intracellular signals that activate integrin αL (“inside-out” signaling). This results in the conformational change of integrin αL, which increases its affinity for its ligand ICAM-1,-2, and -3. Furthermore, this signaling releases integrin αL from the actin cytoskeleton, resulting in clustering of this surface molecule. The high affinity conformation and the clustering of integrin αL increases the adhesion between it and ICAM. This adhesion is essential for leukocyte activation and results in “outside-in” signaling which induces the polymerization of actin, thereby stabilizing the integrin αL-ICAM interaction. This interaction is necessary for CTL and NK mediated killing, T and B cell responses, ADCC, and immune cell extravasation.

Immunofluorescence, Immunoprecipitation, Western Blotting

Spezifikation

Spezifikation

CD11a
Monoclonal
250μg/mL
Integrin αL; LFA-1α
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
27
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human LFA-1α aa. 64-199
50 μg
Immunology
Human
IgG1 κ
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.