Learn More
CD14 Mouse anti-Human, Brilliant Violet 650, Clone: M5E2, BD
Mouse Monoclonal Antibody
330.00 € - 680.00 €
Spezifikation
Antigen | CD14 |
---|---|
Klon | M5E2 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Brilliant Violet 650 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15812779
|
BD Biosciences
563420 |
25 Tests |
330.00 €
1 Stück |
Verfügbar ab: 15-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15802779
|
BD Biosciences
563419 |
100 Tests |
680.00 €
1 Stück |
Verfügbar ab: 15-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will bespillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Spezifikation
CD14 | |
Flow Cytometry | |
Brilliant Violet 650 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein |
M5E2 | |
Monoclonal | |
Mouse | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.