missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, V500, Clone: 3G8, BD

Mouse Monoclonal Antibody

316.00 € - 665.00 €

Spécification

Antigen CD16
Klon 3G8
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Konjugat Violet Fluor 500 (V500)
Consulter d'autres spécifications

Produits 2
Code produit Marque Menge Prix Quantité et disponibilité  
Code produit Marque Menge Prix Quantité et disponibilité  
15801009
missing translation for 'viewDocuments' Détails de la promotion
BD Biosciences
561393
25 Tests
316.00 €
1 Stück
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
15811009
missing translation for 'viewDocuments' Détails de la promotion
BD Biosciences
561394
100 Tests
665.00 €
1 Stück
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

The 3G8 monoclonal antibody specifically binds to the 50-65 kDa transmembrane form of the IgG Fc Receptor (FcγRIII), a human NK cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. The 3G8 antibody blocks the binding of soluble immune complexes to granulocytes. The 3G8 antibody is reported (Vossebeld et al., 1997) to increase intracellular calcium levels in human neutrophils by interacting with bothFcγRIIa and FcγRIIIb molecules. This antibody has also been reported to induce homotypic neutrophil aggregation.

The antibody is conjugated to BD Horizon™ V500, which has been developed for use in multicolor flow cytometry experiments and is

available exclusively from BD Biosciences. It is excited by the Violet laser with an Ex max of 415nm and Em Max at 500 nm. BD Horizon V500 conjugates emit at a similar wavelength to Amcyan yet exhibit reduced spillover into the FITC channel. For more information on BD Horizon V500, visit bdbiosciences.com/colors.

When compensating dyes in this spectral range (such as Horizon™ V500 and AmCyan), the most accurate compensation can be obtained using single stained cellular controls. Due to spectral differences between cells and beadsin this channel, using BD CompBeads can result in spillover errors for V500 and AmCyan reagents.Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended.Different V500 reagents (e.g. CD4 vs. CD45) can have slightly different fluorescence spillover therefore, it may also be necessary to use clone specific compensation controls when using these reagents.

Spécification

Spécification

CD16
Flow Cytometry
Violet Fluor 500 (V500)
RUO
Aqueous buffered solution containing protein stabilizer, glycerol and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
3G8
Monoclonal
Mouse
Baboon, Cynomolgus, Human, Rhesus Monkey
Human polymorphonuclear leukocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Vidéos
FDS
Documents

Documents

Certificats

For Research Use Only.