missing translation for 'onlineSavingsMsg'
Learn More

CD19 Rat anti-Mouse, PerCP-Cy5.5, Clone: 1D3, BD

Rat Monoclonal Antibody

121.00 € - 394.00 €

Spezifikation

Antigen CD19
Isotyp IgG2a, κ
Immunogen Mouse CD19 Transfected Cell Line
Rezeptur Aqueous buffered solution containing ≤0.09% sodium azide.
Konjugat PerCP-Cyanine5.5
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15800789
Unterlagen ansehen
BD Biosciences
561113
25μg 121.00 €
1 Stück Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab
10-11-2020
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
15876468
Unterlagen ansehen
BD Biosciences
551001
0.1mg 394.00 €
1 Stück Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab
10-11-2020
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung

Beschreibung

The 1D3 antibody reacts with CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 95-kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell throughthe mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.
Spezifikation

Spezifikation

CD19
Mouse CD19 Transfected Cell Line
PerCP-Cyanine5.5
Immunology
Rat
Affinity Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG2a, κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
1D3
0.2mg/mL

For Research Use Only.