missing translation for 'onlineSavingsMsg'
Learn More

CD195 Mouse anti-Human, PE-Cy5, Clone: 2D7/CCR5, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 556889

Weitere Details : Netto-Gewicht : 0.02000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15898788

  • 805.98 € / 1 Stück
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The 2D7/CCR5 monoclonal antibody specifically binds to the human chemokine receptor, CCR5. CCR5 is also known as CD195. CCR5 is a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the beta chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+CD45RO+CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: Regulated upon Activation Normal T-cell Expressed and Secreted (RANTES), Macrophage Inflammatory Protein-1 (MIP-1) and Monocyte Chemoattractant Protein 2 (MCP-2). Additionally, CCR5 has been found to be a coreceptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection.

Spezifikation

Spezifikation

CD195
Monoclonal
PE-Cyanine5
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Human CCR5 Transfected Cell Line
100 Tests
Immunology
Human
IgG2a κ
Flow Cytometry
2D7/CCR5
CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.