Learn More
CD23/Fc epsilon RII Mouse anti-Human, Clone: 138633, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1231
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418475
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD23/Fc epsilon RII Monoclonal antibody MAB1231 Specifically detects CD23/Fc epsilon RII in Human samples. It is validated for ELISA Capture
Spezifikation
Spezifikation
CD23/Fc epsilon RII
138633
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FCER2
Mouse myeloma cell line NS0-derived recombinant human CD23 Met150-Ser321 Accession # P06734
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2208
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P06734
BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD23 in ELISAs and Western blots.