Learn More
CD24 Monoclonal Antibody (YBM5.10.4), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517892
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17892 targets CD24/HSA in ELISA, FACS, and WB applications and shows reactivity with Mouse samples. The MA5-17892 immunogen is mouse monoclonal antibodies raised against mouse CD24.
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for ELISA, Flow Cytometry, Western Blot.Spezifikation
CD24 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P24807 | |
Cd24a | |
Mouse monoclonal antibodies raised against mouse CD24. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2c |
ELISA, Flow Cytometry, Western Blot | |
YBM5.10.4 | |
Unconjugated | |
Cd24a | |
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen | |
Rat | |
Protein G | |
RUO | |
12484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |