missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dpp4 Rat anti-Mouse, PerCP-Cy5.5, Clone: H194-112 (H194-112.12.4.4.5), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen A15866
Weitere Details : Netto-Gewicht : 0.00280kg
Spezifikation
Dpp4 | |
Monoclonal | |
PerCP-Cyanine5.5 | |
RP23-87G8.2, Cd26, Dpp-4, THAM | |
25 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry | |
H194-112 (H194-112.12.4.4.5) | |
PBS, pH 7.2, with 0.1% gelatin and 0.09% sodium azide | |
Rat | |
RUO | |
13482 | |
Store at 2 - 8 °C. | |
IgG2a |