missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Monoclonal Antibody (DEN-3), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA112073
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD27 | |
Monoclonal | |
Unconjugated | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
Protein A/G | |
RUO | |
939 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
DEN-3 | |
PBS with 0.08% sodium azide | |
P26842 | |
CD27 | |
White blood cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |