Learn More
CD27 Monoclonal Antibody (CLB-27/1), Qdot™ 605, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen Q10065
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Qdot™ Antibody (Ab) conjugates possess a bright fluorescence emission that makes them well suited for the detection of low-abundance extracellular proteins. Approximately the same size as R-phycoerythrin (R-PE) and compatible with existing organic fluorophore conjugates, Qdot™ Ab conjugates can be excited with any wavelength below their emission maximum, but are best excited by UV or violet light. The narrow, symmetric emission profiles of Qdot Ab conjugates allow for minimal compensation when using a single excitation source, and the very long stoke shifts enable better, more efficient multicolor assays using the 405 nm violet laser. Available in multiple colors for use in flow cytometry, these advantages make Qdot Ab conjugates powerful tools for antibody labeling and staining. Staining: Stain cells in any standard staining buffer, such as phosphate buffered saline (PBS) with 1% bovine serum albumin (BSA). Use 1 L of Qdot antibody conjugate per 1 x 10^6 cells in a 100 μL staining volume (20 nM final concentration of Qdot™ ocrystals). Qdot™ ocrystal conjugates may be mixed with other antibodies, but use the diluted conjugates on the day of dilution. Qdot ocrystal conjugates can be used for surface staining applications with most conventional sample preparation reagents, such as Cal-Lyse™ and FIX & PERM™ reagents, with minimal affect on fluorescence.
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD27 | |
Monoclonal | |
Qdot 605 | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
purified | |
RUO | |
939 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
CLB-27/1 | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
P26842 | |
CD27 | |
Human CD27. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |