Learn More
ITGB1 Monoclonal Antibody (4B7R), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA182003
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD29 monoclonal antibody, clone 4B7R recognizes the human integrin beta 1 subunit, also known as CD29, a approximately 130 kDa (red) 115 kDa (non-red) single pass type I transmembrane glycoprotein expressed by most leucocytes and mesenchymal stem cells.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human samples. It is validated for Flow Cytometry.Spezifikation
ITGB1 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P05556 | |
ITGB1 | |
Ocular melanoma cell line V+B2. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
4B7R | |
PE | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein A | |
RUO | |
3688 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |