Learn More
CD3 Mouse anti-Rat, APC, Clone: 1F4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 557030
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
The 1F4 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes, peripheral T lymphocytes, and dendritic epidermal T cells. In vitro, immobilized 1F4 mAb induces T-cell proliferation, and soluble mAb synergizes with PMA for T-cell activation. Pre-incubation of splenocytes with mAb 1F4 blocks staining with mAb G4.18 (Cat. No. 559974/554830).
Spezifikation
CD3 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
F344 rat splenic T cells stimulated with PMA plus A23187 | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
1F4 | |
APC | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgM κ |
For Research Use Only.