missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 Monoclonal Antibody (Ber-H2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA126282
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-26282 detects CD30 from human samples.
CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
CD30 | |
Monoclonal | |
Not Determined | |
tissue culture supernatant with 0.05% sodium azide | |
P28908 | |
TNFRSF8 | |
Co cell line established from a patient with Hodgkins disease of T-cell lineage. | |
6 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
Ber-H2 | |
Unconjugated | |
TNFRSF8 | |
Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30 | |
Mouse | |
purified | |
RUO | |
943 | |
4° C, do not freeze | |
Liquid |