missing translation for 'onlineSavingsMsg'
Learn More

CD30 Monoclonal Antibody (CON6D/ C2), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA512632

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11334683

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

MA5-12632 targets CD30 in IHC (P), WB and FACS applications and shows reactivity with Human samples. The MA5-12632 immunogen is hodgkin's lymphoma cell line L428.

CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

CD30
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P28908
TNFRSF8
Hodgkin's lymphoma cell line L428
500 μL
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
CON6D/ C2
Unconjugated
TNFRSF8
Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30
Mouse
Protein G
RUO
943
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote