Learn More
CD30 Ligand/TNFSF8 Mouse anti-Human, Clone: 116615, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB774
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411856
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD30 Ligand/TNFSF8 Monoclonal antibody MAB774 Specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
CD30 Ligand/TNFSF8
116615
Western Blot 1 ug/mL
P32971
CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD30 Ligand/TNFSF8 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse CD30 Ligand is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFSF8
Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8 Gln63-Asp234 Accession # P32971
Protein A or G purified from ascites
RUO
Primary
944
Reconstitute at 0.5 mg/mL in sterile PBS.