missing translation for 'onlineSavingsMsg'
Learn More

CD153 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA547045

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15597532

  • 461.00 € / 100 Mikrogramm
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) CD27 Ligand is observed and less than 1% cross-reactivity with rhCD40 Ligand is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

CD153 Polyclonal antibody specifically detects CD153 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

CD153
Polyclonal
Unconjugated
Tnfsf8
CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8
Goat
Antigen affinity chromatography
RUO
944
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P32971
Tnfsf8
Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand Gln63-Asp234
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote