Learn More
CD45 Mouse, APC, Clone: D058-1283, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561290
Weitere Details : Netto-Gewicht : 0.01000kg
Description
D058-1283 is a CD45 monoclonal antibody specific for non-human primate leucocytes, developed using Rhesus peripheral whole blood as immunogen. It does not cross react with human leucocytes. This antibody reacts with Baboon, Rhesus and Cynomolgus Macaque leucocytes in a similar pattern as seen with CD45 binding to the Leukocyte Common Antigen on human cells. Immunophenotypic analysis shows that D058-1283 binds to lymphocytes, monocytes and granulocytes of non-human primate blood samples. This antibody is able to block the binding of monoclonal antibody TÜ116; a reported anti-human CD45 antibody that cross-reacts with nonhuman primate leucocytes. In Western blot analysis, the D058-1283 antibody identifies a band of approximately 180-200 kDa molecular weight .
Specifications
CD45 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rhesus peripheral whole blood | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
D058-1283 | |
Pan Leukocyte, NHP-specific | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |