missing translation for 'onlineSavingsMsg'
Learn More

CD45RA Mouse anti-Human, PE-Cy5, Clone: HI100, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561885

Weitere Details : Netto-Gewicht : 0.02000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15801439

  • 198.00 € / 1 Stück

Verfügbar ab
16-10-2020

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.

Spezifikation

Spezifikation

CD45RA
HI100
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE-Cyanine5
Affinity Purified
IgG2b, κ
RUO
Primary
Human
Videos
Dokumentation

For Research Use Only.