Learn More
CD5 Rat anti-Mouse, Biotin, Clone: 53-7.3, BD
Rat Monoclonal Antibody
203.00 € - 750.00 €
Spezifikation
Antigen | CD5 |
---|---|
Klon | 53-7.3 |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15896928
|
BD Biosciences
553018 |
0.1 mg |
203.00 €
1 Stück |
Verfügbar ab: 15-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15806938
|
BD Biosciences
553019 |
0.5 mg |
750.00 €
1 Stück |
Verfügbar ab: 15-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The 53-7.3 clone has been reported to react with a monomorphic epitope of CD5, a member of the scavenger receptor cysteine-rich protein superfamily and the major ligand of CD72, found on thymocytes, T lymphocytes, thymic NK-T cells, and a subset of B lymphocytes, but not on NK cells or splenic NK-T cells. The level of surface CD5 expression is developmentally regulated in the thymus, starting with low levels on CD4-CD8- thymocytes and increasing as they mature to CD4+CD8+ then CD4+CD8- or CD4-CD8+ thymocytes. Relatively high levels are maintained on peripheral T lymphocytes. The level of CD5 antigen detected on T helper cells has been reported to be somewhat higher than that on T cytotoxic/suppressor and B cells. Few, if any, intestinal intraepithelial lymphocytes bearing the γδ T-cell receptor express CD5. Phenotypic, anatomical, functional, developmental, and pathogenic characteristics of peripheral CD5+ B cells suggest that they may represent a distinct lineage, known as B-1 cells. The frequency of these CD5+ B cells has been reported to show strain-dependent variation. An additional population of CD5+ B lymphocytes resides in the thymus, where it matures from intrathymic B-cell progenitors. It has been proposed that CD5 is a costimulatory molecule which mediates interactions of cells in the immune system and negatively regulates signal transduction mediated by the T-cell receptor and B-cell receptor. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 53-7.3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus / Spleen
Flow Cytometry
Spezifikation
CD5 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Lymphocyte antigen 1; Cd5; Ly-12; Ly-A; Lyt-1 |
53-7.3 | |
Flow Cytometry | |
Biotin | |
RUO | |
Mouse | |
Mouse Thymus / Spleen | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.