Learn More
CD6 Mouse anti-Human, Clone: 123119, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB627
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411296
  • 420.42 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 22-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
CD6 Monoclonal antibody MAB627 Specifically detects CD6 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
CD6
123119
Western Blot 1 ug/mL
CAA43306
CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD6 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD6
Mouse myeloma cell line NS0-derived recombinant human CD6 His18-Glu398 Accession # CAA43306
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
923
Reconstitute at 0.5 mg/mL in sterile PBS.