Learn More
CD63 Mouse anti-Human, Unlabeled, Clone: H5C6, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 556019
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The H5C6 monoclonal antibody reacts with CD63. CD63 is a 53 kDa, type III lysosomal glycoprotein, expressed on activated platelets, monocytes and macrophages. This molecule is also referred to in the literature as LIMP, gp55, melanoma-associated antigen ME491, Pltgp40, LAMP-3 and is a member of the tetraspan transmembrane 4 superfamily (TM4SF). It is widely expressed on surface and in the cytoplasm of various hematopoietic (monocytes, macrophages) and non-hematopoietic cells (endothelium, fibroblasts, osteoclasts, smooth muscle).
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Spezifikation
CD63 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Splenic Adherent Cells | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
H5C6 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.