Learn More
CD8 Mouse anti-Human, APC-H7, Clone: SK1, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560179
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD8 recognizes the 32kDa a-subunit of a disulfide-linked bimolecular complex. The majority of peripheral blood CD8+ T lymphocytes express an a/b heterodimer (Mr 32, 30kDa), while CD8+CD16+ natural killer (NK) lymphocytes and CD8+ T-cell receptor (TCR)-γ/δ+ lymphocytes express a/a homodimer (Mr 30kDa). CD8+TCR-α/β+ lymphocytes can express either an α/α homodimer or α/β heterodimer. The CD8 antigenic determinant binds to class I major histocompatibility (MHC) molecules resulting in increased adhesion between the CD8+ T lymphocytes and target cells. Binding of the CD8 antigen is coupled to a protein tyrosine kinase p56lck. The CD8:p56lck complex can play a role in T-lymphocyte activation through mediation of the interactions between the CD8 antigen and the CD3/TCR complex.
Host Species: Mouse
Clone: SK1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Peripheral Blood T Cells
Flow Cytometry
Spezifikation
CD8 | |
Monoclonal | |
APC-H7 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human Peripheral Blood T Cells | |
100 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
SK1 | |
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.