missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Monoclonal Antibody (MEM-233), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA119590
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD80 Monoclonal antibody specifically detects CD80 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD80 | |
Monoclonal | |
FITC | |
CD80 | |
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
941 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-233 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P33681 | |
CD80 | |
Extracellular domain of human CD80 fused to human IgG1(Fc). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |